Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gata6
Entrez Gene ID
58076
Gene Name
GATA binding protein 6
Gene Aliases
GATA-5, cb603
Chromosome Location
Chr.2: 4207028 - 4217877 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017358958.2 XP_017214447.1 6 - 7 1586 116
GenBank mRNA AF191578.1 - 5 - 6 1036 116

View Full Product Details