Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aldocb
Entrez Gene ID
369193
Gene Name
aldolase C, fructose-bisphosphate, b
Gene Aliases
aldoc
Chromosome Location
Chr.21: 39609482 - 39639954 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194384.1 NP_919365.1 4 - 5 446 133
GenBank mRNA AF533647.1 - 4 - 5 452 133
BC053192.1 - 4 - 5 446 133
BC164189.1 - 3 - 4 413 133

View Full Product Details