Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdia6
Entrez Gene ID
322160
Gene Name
protein disulfide isomerase family A, member 6
Gene Aliases
pdi-p5, pdip5, wu:fb51h08, wu:fc09e09
Chromosome Location
Chr.20: 31217591 - 31231122 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197933.2 NP_922915.2 8 - 9 862 111
GenBank mRNA AF387900.1 - 8 - 9 909 111
BC044507.1 - 8 - 9 862 111
BC165811.1 - 8 - 9 870 111

View Full Product Details