Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
drd2l
Entrez Gene ID
378718
Gene Name
dopamine receptor D2 like
Gene Aliases
drd2b
Chromosome Location
Chr.16: 54248542 - 54296239 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197935.1 NP_922917.1 3 - 4 557 66
XM_009292744.3 XP_009291019.1 4 - 5 929 66
GenBank mRNA AY333791.1 - 3 - 4 557 66

View Full Product Details