Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
figla
Entrez Gene ID
387259
Gene Name
folliculogenesis specific bHLH transcription factor
Gene Aliases
FIGalpha, fig1a, zgc:109752
Chromosome Location
Chr.5: 9195 - 18175 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198919.2 NP_944601.2 3 - 4 617 85
GenBank mRNA AY302567.1 - 3 - 4 471 85
BC092676.1 - 3 - 4 617 85
BC164317.1 - 3 - 4 505 85
EU723768.1 - 3 - 4 563 85

View Full Product Details