Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
itm2ba
Entrez Gene ID
323632
Gene Name
integral membrane protein 2Ba
Gene Aliases
itm2b, wu:fc05f02, zgc:55770, zgc:85688
Chromosome Location
Chr.6: 10015525 - 10026822 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199646.1 NP_955940.1 5 - 6 784 70
GenBank mRNA BC044480.1 - 5 - 6 784 70
BC068357.1 - 5 - 6 782 70
BC128819.1 - 5 - 6 760 70
BC164594.1 - 5 - 6 729 70

View Full Product Details