Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atf1
Entrez Gene ID
325609
Gene Name
activating transcription factor 1
Gene Aliases
wu:fc92c07, zgc:64099
Chromosome Location
Chr.6: 78083 - 91525 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199723.1 NP_956017.1 4 - 5 472 84
GenBank mRNA BC053252.1 - 4 - 5 472 84

View Full Product Details