Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan31
Entrez Gene ID
393251
Gene Name
tetraspanin 31
Gene Aliases
sas, zgc:56710
Chromosome Location
Chr.23: 36730750 - 36750304 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200281.1 NP_956575.1 4 - 5 495 80
XM_021469763.1 XP_021325438.1 4 - 5 445 80
GenBank mRNA AY398411.1 - 4 - 5 532 80
BC049345.1 - 4 - 5 495 80

View Full Product Details