Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
matn3b
Entrez Gene ID
497645
Gene Name
matrilin 3b
Gene Aliases
zgc:123028
Chromosome Location
Chr.13: 32627283 - 32635905 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012385.1 NP_001012385.1 2 - 3 624 104
XM_005156825.4 XP_005156882.1 2 - 3 670 104
GenBank mRNA AJ698458.1 - 2 - 3 624 104
AJ717297.1 - 2 - 3 624 104

View Full Product Details