Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
olfcw1
Entrez Gene ID
553200
Gene Name
olfactory receptor C family, w1
Gene Aliases
si:ch211-128b10.3, v2r5, v2rd1, zgc:112465
Chromosome Location
Chr.18: 181102 - 185628 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018147.1 NP_001018157.1 5 - 6 1735 80
GenBank mRNA BC095823.1 - 5 - 6 1735 80

View Full Product Details