Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cfap52
Entrez Gene ID
553666
Gene Name
cilia and flagella associated protein 52
Gene Aliases
fj15f10, si:dkey-28k21.1, wdr16, wu:fj15f10, zgc:111988
Chromosome Location
Chr.6: 312137 - 328005 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020639.1 NP_001018475.1 4 - 5 563 99
GenBank mRNA BC095635.1 - 4 - 5 563 99

View Full Product Details