Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfap2d
Entrez Gene ID
566838
Gene Name
transcription factor AP-2 delta (activating enhancer binding protein 2 delta)
Gene Aliases
si:dkey-5e16.2, zgc:114196
Chromosome Location
Chr.20: 47717840 - 47732703 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025546.1 NP_001020717.1 2 - 3 760 94
XM_005160887.4 XP_005160944.1 2 - 3 1038 94
GenBank mRNA BC097244.1 - 2 - 3 760 94

View Full Product Details