Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hapln4
Entrez Gene ID
559818
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
si:dkey-19f23.2
Chromosome Location
Chr.22: 17822639 - 17831253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044907.1 NP_001038372.1 2 - 3 584 75
XM_005170459.4 XP_005170516.1 3 - 4 794 75
GenBank mRNA BC142773.1 - 1 - 2 656 75

View Full Product Details