Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sin3aa
Entrez Gene ID
563090
Gene Name
SIN3 transcription regulator family member Aa
Gene Aliases
zgc:162128
Chromosome Location
Chr.25: 6349896 - 6391307 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098180.1 NP_001091650.1 20 - 21 3741 86
XM_005162970.4 XP_005163027.1 20 - 21 3700 86
GenBank mRNA BC139856.1 - 20 - 21 3741 86

View Full Product Details