Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddx4
Entrez Gene ID
30263
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 4
Gene Aliases
vas, vasa, vlg, wu:fi24g05, zgc:109812, zgc:158535
Chromosome Location
Chr.10: 6884126 - 6905588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131057.1 NP_571132.1 19 - 20 1055 73
XM_005156453.4 XP_005156510.1 19 - 20 1067 73
XM_017357926.2 XP_017213415.1 19 - 20 1064 73
XM_017357927.2 XP_017213416.1 18 - 19 1034 73
XM_021479102.1 XP_021334777.1 18 - 19 1031 73
XM_021479103.1 XP_021334778.1 17 - 18 934 73
XM_021479104.1 XP_021334779.1 17 - 18 978 73
XM_021479105.1 XP_021334780.1 16 - 17 898 73
GenBank mRNA AB005147.1 - 19 - 20 1055 73
BC129275.1 - 17 - 18 73
Y12007.1 - - 972 73

View Full Product Details