Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddx4
Entrez Gene ID
30263
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 4
Gene Aliases
vas, vasa, vlg, wu:fi24g05, zgc:109812, zgc:158535
Chromosome Location
Chr.10: 6884126 - 6905588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131057.1 NP_571132.1 22 - 23 1513 138
XM_005156453.4 XP_005156510.1 22 - 23 1525 138
XM_017357926.2 XP_017213415.1 22 - 23 1522 138
XM_017357927.2 XP_017213416.1 21 - 22 1492 138
XM_021479102.1 XP_021334777.1 21 - 22 1489 138
XM_021479103.1 XP_021334778.1 20 - 21 1392 138
XM_021479104.1 XP_021334779.1 20 - 21 1436 138
XM_021479105.1 XP_021334780.1 19 - 20 1356 138
GenBank mRNA AB005147.1 - 22 - 23 1513 138
BC129275.1 - 20 - 21 1444 138
Y12007.1 - - 1430 138

View Full Product Details