Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddx4
Entrez Gene ID
30263
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 4
Gene Aliases
vas, vasa, vlg, wu:fi24g05, zgc:109812, zgc:158535
Chromosome Location
Chr.10: 6884126 - 6905588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131057.1 NP_571132.1 6 - 7 255 62
XM_005156453.4 XP_005156510.1 6 - 7 270 62
XM_017357926.2 XP_017213415.1 6 - 7 270 62
XM_017357927.2 XP_017213416.1 6 - 7 270 62
XM_021479102.1 XP_021334777.1 5 - 6 231 62
XM_021479103.1 XP_021334778.1 4 - 5 137 62
XM_021479104.1 XP_021334779.1 5 - 6 214 62
XM_021479105.1 XP_021334780.1 4 - 5 137 62
GenBank mRNA AB005147.1 - 6 - 7 255 62
BC095001.1 - - 211 62
BC129275.1 - 5 - 6 222 62

View Full Product Details