Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ddx4
Entrez Gene ID
30263
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 4
Gene Aliases
vas, vasa, vlg, wu:fi24g05, zgc:109812, zgc:158535
Chromosome Location
Chr.10: 6884126 - 6905588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131057.1 NP_571132.1 7 - 8 304 113
XM_005156453.4 XP_005156510.1 7 - 8 319 113
XM_017357926.2 XP_017213415.1 7 - 8 319 113
XM_017357927.2 XP_017213416.1 7 - 8 319 113
XM_021479102.1 XP_021334777.1 6 - 7 280 113
XM_021479103.1 XP_021334778.1 5 - 6 186 113
XM_021479104.1 XP_021334779.1 6 - 7 263 113
XM_021479105.1 XP_021334780.1 5 - 6 186 113
GenBank mRNA AB005147.1 - 7 - 8 304 113
BC129275.1 - 6 - 7 271 113

View Full Product Details