Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmo1
Entrez Gene ID
280646
Gene Name
LIM domain only 1
Gene Aliases
cb524, etID41074.7, lmo1b, wu:fb76g02, zgc:109794
Chromosome Location
Chr.7: 28109362 - 28148312 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328502.1 NP_001315431.1 3 - 4 859 68
NM_173219.3 NP_775326.1 2 - 3 799 68
XM_021477388.1 XP_021333063.1 3 - 4 558 68
XM_021477389.1 XP_021333064.1 2 - 3 474 68
GenBank mRNA AF398514.1 - 2 - 3 755 68
BC092690.1 - 2 - 3 766 68
BC164905.1 - 2 - 3 267 68
EH453517.1 - 3 - 4 411 68

View Full Product Details