Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arntl1b
Entrez Gene ID
352935
Gene Name
aryl hydrocarbon receptor nuclear translocator-like 1b
Gene Aliases
bmal1b, zfBMAL3
Chromosome Location
Chr.7: 209733 - 240518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178300.1 NP_840085.1 15 - 16 1611 85
XM_005166616.4 XP_005166673.1 18 - 19 85
XM_009303573.3 XP_009301848.1 17 - 18 85
XM_021477412.1 XP_021333087.1 17 - 18 85
GenBank mRNA AB087257.1 - 15 - 16 1611 85
BC134895.1 - 18 - 19 85

View Full Product Details