Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mmp14a
Entrez Gene ID
373882
Gene Name
matrix metallopeptidase 14a (membrane-inserted)
Gene Aliases
cb386, sb:cb386, wu:fb72b09
Chromosome Location
Chr.7: 24124298 - 24149631 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194416.1 NP_919397.1 1 - 2 411 95
GenBank mRNA AY324196.1 - 1 - 2 411 95
BC131864.1 - 1 - 2 403 95

View Full Product Details