Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
esf1
Entrez Gene ID
326718
Gene Name
ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae)
Gene Aliases
fe06g12, im:7138472, wu:fe06g12, zgc:66241
Chromosome Location
Chr.1: 51202984 - 51219965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199745.2 NP_956039.2 4 - 5 908 67
GenBank mRNA BC054666.1 - - 544 67
BC153482.1 - 2 - 3 532 67

View Full Product Details