Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vil1
Entrez Gene ID
393207
Gene Name
villin 1
Gene Aliases
vil1l, zgc:55779
Chromosome Location
Chr.9: 44910768 - 44953424 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200238.1 NP_956532.1 4 - 5 426 67
XM_005167920.4 XP_005167977.1 4 - 5 504 67
GenBank mRNA BC047186.1 - 4 - 5 426 67
BC165692.1 - 3 - 4 379 67

View Full Product Details