Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab3aa
Entrez Gene ID
445024
Gene Name
RAB3A, member RAS oncogene family, a
Gene Aliases
rab3a, zgc:92276
Chromosome Location
Chr.2: 280020 - 290608 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003419.1 NP_001003419.1 4 - 5 487 67
XM_009298545.3 XP_009296820.1 4 - 5 579 67
GenBank mRNA BC075980.1 - 4 - 5 487 67

View Full Product Details