Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lhfpl6
Entrez Gene ID
494110
Gene Name
LHFPL tetraspan subfamily member 6
Gene Aliases
lhfp, zgc:101683
Chromosome Location
Chr.10: 31951338 - 32038263 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008653.1 NP_001008653.1 2 - 3 700 81
GenBank mRNA BC086720.1 - 2 - 3 700 81
BC165936.1 - 1 - 2 419 81

View Full Product Details