Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
xrcc3
Entrez Gene ID
541414
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 3
Gene Aliases
im:7142103, si:dkey-11b8.1, zgc:101608
Chromosome Location
Chr.20: 20896690 - 20901329 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013541.1 NP_001013559.1 6 - 7 924 71
GenBank mRNA BC090820.1 - 6 - 7 924 71
BC164394.1 - 6 - 7 859 71

View Full Product Details