Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kin
Entrez Gene ID
555316
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
fc49f10, si:ch211-235a22.3, wu:fc49f10, zgc:123169
Chromosome Location
Chr.4: 25181572 - 25188103 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030086.2 NP_001025257.2 6 - 7 859 132
GenBank mRNA BC109449.1 - 6 - 7 859 132

View Full Product Details