Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prtga
Entrez Gene ID
563834
Gene Name
protogenin homolog a (Gallus gallus)
Gene Aliases
-
Chromosome Location
Chr.Unknown: 4017 - 28442 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045030.1 - 696 64
GenBank mRNA DQ360115.1 - - 696 64

View Full Product Details