Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sppl2
Entrez Gene ID
325540
Gene Name
signal peptide peptidase-like 2
Gene Aliases
SPPL2B, wu:fc16e01, wu:fc85d12, zgc:136525
Chromosome Location
Chr.2: 37480654 - 37503010 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045244.2 NP_001038709.2 8 - 9 1018 91
XM_021478906.1 XP_021334581.1 8 - 9 1013 91
XM_021478911.1 XP_021334586.1 8 - 9 1013 91
GenBank mRNA AY926457.1 - 8 - 9 1002 91
BC044512.1 - 8 - 9 986 91
BC116574.1 - 8 - 9 1003 91
EH571140.1 - 6 - 7 644 91

View Full Product Details