Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
erbin
Entrez Gene ID
566811
Gene Name
erbb2 interacting protein
Gene Aliases
erbb2ip, zgc:152984
Chromosome Location
Chr.10: 15454598 - 15599149 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193543.1 NP_001180472.1 14 - 15 1766 60
XM_005155576.4 XP_005155633.1 14 - 15 2024 60
XM_005155577.3 XP_005155634.1 14 - 15 1766 60
XM_005155578.4 XP_005155635.1 14 - 15 2020 60
GenBank mRNA BC125845.1 - 13 - 14 1679 60
BC146713.1 - 14 - 15 1766 60
BC171620.1 - 14 - 15 1664 60

View Full Product Details