Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mylk2
Entrez Gene ID
558685
Gene Name
myosin light chain kinase 2
Gene Aliases
mlck2
Chromosome Location
Chr.23: 32044422 - 32060883 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110520.2 NP_001103990.1 6 - 7 1269 70

View Full Product Details