The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130959.2 | NP_571034.1 | 2 - 3 | 131 | 94 |
GenBank mRNA | KM655728.1 | - | 2 - 3 | 102 | 94 |
U10870.1 | - | 2 - 3 | 135 | 94 |