Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
elavl3
Entrez Gene ID
30732
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HuC, elrc, id:ibd1248, wu:fb77b03, zHuC
Chromosome Location
Chr.3: 46798511 - 46817871 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131449.1 NP_571524.1 3 - 4 363 99
XM_005164218.4 XP_005164275.1 3 - 4 736 99
XM_005164219.4 XP_005164276.1 3 - 4 736 99
XM_005164220.4 XP_005164277.1 3 - 4 736 99
XM_009299733.3 XP_009298008.1 3 - 4 736 99
XM_017353188.2 XP_017208677.1 3 - 4 736 99
XM_017353190.2 XP_017208679.1 3 - 4 736 99
XM_017353191.2 XP_017208680.1 3 - 4 736 99
GenBank mRNA BC065343.1 - 3 - 4 703 99
U62018.1 - 3 - 4 363 99

View Full Product Details