The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309399.1 | NP_001296328.1 | 4 - 5 | 1067 | 86 |
NM_001309400.1 | NP_001296329.1 | 4 - 5 | 1067 | 86 | |
NM_152962.3 | NP_694494.2 | 4 - 5 | 1067 | 86 | |
XM_009304303.3 | XP_009302578.1 | 4 - 5 | 1109 | 86 | |
XM_009304304.3 | XP_009302579.1 | 4 - 5 | 1110 | 86 | |
GenBank mRNA | AF389400.2 | - | 3 - 4 | 86 | |
AY197498.1 | - | 3 - 4 | 86 | ||
AY197499.1 | - | 3 - 4 | 86 | ||
AY197500.1 | - | 3 - 4 | 86 | ||
AY197501.1 | - | 3 - 4 | 389 | 86 | |
BC162342.1 | - | 3 - 4 | 86 | ||
CU458752.1 | - | 3 - 4 | 86 |