Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbx15
Entrez Gene ID
246222
Gene Name
T-box transcription factor 15
Gene Aliases
zgc:158205, zgc:63576
Chromosome Location
Chr.9: 21034870 - 21067986 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF448504.1 - 6 - 7 1190 74

View Full Product Details