Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mknk2b
Entrez Gene ID
373121
Gene Name
MAPK interacting serine/threonine kinase 2b
Gene Aliases
mknk2, wu:fb37e05, wz5090
Chromosome Location
Chr.8: 20438889 - 20449541 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194402.1 NP_919383.1 3 - 4 369 72
GenBank mRNA BC044375.1 - 3 - 4 369 72
BC165746.1 - 2 - 3 173 72

View Full Product Details