Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prkcbp1l
Entrez Gene ID
323479
Gene Name
protein kinase C binding protein 1, like
Gene Aliases
im:7138336, wu:fb99d12, zgc:63775
Chromosome Location
Chr.23: 9809630 - 9863632 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199641.1 NP_955935.1 17 - 18 2980 88
XM_005161943.4 XP_005162000.1 18 - 19 3089 88
XM_005161944.4 XP_005162001.1 18 - 19 3551 88
XM_005161945.4 XP_005162002.1 18 - 19 3394 88
XM_005161946.4 XP_005162003.1 18 - 19 3010 88
XM_009296662.3 XP_009294937.1 18 - 19 2989 88
XM_009296664.3 XP_009294939.1 18 - 19 2992 88
XM_009296665.3 XP_009294940.1 18 - 19 3089 88
GenBank mRNA BC058064.1 - 17 - 18 2980 88

View Full Product Details