Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prkcbp1l
Entrez Gene ID
323479
Gene Name
protein kinase C binding protein 1, like
Gene Aliases
im:7138336, wu:fb99d12, zgc:63775
Chromosome Location
Chr.23: 9809630 - 9863632 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199641.1 NP_955935.1 2 - 3 218 94
XM_005161943.4 XP_005162000.1 3 - 4 327 94
XM_005161944.4 XP_005162001.1 3 - 4 789 94
XM_005161945.4 XP_005162002.1 3 - 4 632 94
XM_009296662.3 XP_009294937.1 3 - 4 227 94
XM_009296665.3 XP_009294940.1 3 - 4 327 94
GenBank mRNA BC058064.1 - 2 - 3 218 94
BC091798.1 - 3 - 4 715 94

View Full Product Details