Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl23
Entrez Gene ID
336812
Gene Name
ribosomal protein L23
Gene Aliases
fb06e03, wu:fb06e03, zgc:73149
Chromosome Location
Chr.3: 33423986 - 33427583 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200732.1 NP_957026.1 4 - 5 357 123
GenBank mRNA BC049038.1 - 4 - 5 398 123
BC059509.1 - 4 - 5 357 123

View Full Product Details