Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adh8b
Entrez Gene ID
402841
Gene Name
alcohol dehydrogenase 8b
Gene Aliases
-
Chromosome Location
Chr.13: 12633218 - 12646754 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203460.1 NP_982285.1 6 - 7 736 77
XM_021480549.1 XP_021336224.1 6 - 7 858 77
GenBank mRNA AY309075.1 - 6 - 7 736 77
BC071367.1 - 6 - 7 733 77
BC154296.1 - 5 - 6 591 77

View Full Product Details