Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- wbp11
- Entrez Gene ID
-
402950
- Gene Name
- WW domain binding protein 11
- Gene Aliases
-
zgc:77390
- Chromosome Location
-
Chr.Unknown: 1 - 8488 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205652.1 |
|
- |
1308 |
114 |
GenBank mRNA |
BC056284.1 |
- |
- |
|
114 |
BC065662.1 |
- |
- |
1308 |
114 |
- Gene Symbol
- wbp11
- Entrez Gene ID
-
110437720
- Gene Name
- WW domain binding protein 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 1395484 - 1402828 on Build GRCz11
View Full Product Details