Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hnrnpk
Entrez Gene ID
406267
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
hnrpk, wu:fb37h02, wu:fi34c04, zgc:66162
Chromosome Location
Chr.8: 49745668 - 49766241 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212994.2 NP_998159.2 5 - 6 384 123
XM_005167319.4 XP_005167376.1 5 - 6 383 123
XM_009304280.3 XP_009302555.1 5 - 6 373 123
GenBank mRNA BI704444.1 - 3 - 4 182 123
EB912144.1 - 5 - 6 392 123

View Full Product Details