Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hnrnpk
Entrez Gene ID
406267
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
hnrpk, wu:fb37h02, wu:fi34c04, zgc:66162
Chromosome Location
Chr.8: 49745668 - 49766241 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212994.2 NP_998159.2 9 - 10 1006 100
XM_005167319.4 XP_005167376.1 9 - 10 1005 100
XM_009304280.3 XP_009302555.1 9 - 10 995 100
GenBank mRNA DR719329.1 - 1 - 2 157 100

View Full Product Details