Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnft1
Entrez Gene ID
406509
Gene Name
ring finger protein, transmembrane 1
Gene Aliases
wu:fe23b06, zgc:77306
Chromosome Location
Chr.10: 284506 - 293405 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213228.2 NP_998393.2 5 - 6 954 73
GenBank mRNA BC066378.1 - 5 - 6 925 73

View Full Product Details