Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan7
Entrez Gene ID
406802
Gene Name
tetraspanin 7
Gene Aliases
tm4sf2, wu:fa11g01, wu:fd18e11, wu:fl39g07, zgc:63823
Chromosome Location
Chr.14: 41469478 - 41478369 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214774.2 NP_999939.1 5 - 6 770 92
GenBank mRNA BC055255.1 - 5 - 6 710 92
BC097054.1 - 5 - 6 758 92
BC165114.1 - 5 - 6 642 92

View Full Product Details