Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan7
Entrez Gene ID
406802
Gene Name
tetraspanin 7
Gene Aliases
tm4sf2, wu:fa11g01, wu:fd18e11, wu:fl39g07, zgc:63823
Chromosome Location
Chr.14: 41469478 - 41478369 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214774.2 NP_999939.1 7 - 8 933 87
GenBank mRNA BC055255.1 - 7 - 8 873 87
BC097054.1 - 7 - 8 921 87

View Full Product Details