Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmc1b
Entrez Gene ID
415181
Gene Name
proteasome 26S subunit, ATPase 1b
Gene Aliases
wu:fj14c10, zgc:86923
Chromosome Location
Chr.20: 16721942 - 16740955 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002091.1 NP_001002091.1 7 - 8 717 101
GenBank mRNA BC071538.1 - 7 - 8 717 101

View Full Product Details