Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aagab
Entrez Gene ID
436696
Gene Name
alpha and gamma adaptin binding protein
Gene Aliases
wu:fa20c02, zgc:92663
Chromosome Location
Chr.18: 2601 - 9772 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002423.1 NP_001002423.1 3 - 4 472 68
GenBank mRNA BC076147.1 - 3 - 4 472 68

View Full Product Details