Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aagab
Entrez Gene ID
436696
Gene Name
alpha and gamma adaptin binding protein
Gene Aliases
wu:fa20c02, zgc:92663
Chromosome Location
Chr.18: 2601 - 9772 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002423.1 NP_001002423.1 9 - 10 979 70
GenBank mRNA BC076147.1 - 9 - 10 979 70

View Full Product Details