Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phf20b
Entrez Gene ID
445482
Gene Name
PHD finger protein 20, b
Gene Aliases
zgc:91986
Chromosome Location
Chr.23: 14992800 - 15036822 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004114.1 NP_001004114.1 13 - 14 2006 88
XM_005161993.4 XP_005162050.1 14 - 15 2201 88
GenBank mRNA BC080250.1 - 13 - 14 2006 88

View Full Product Details